3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine

Supplier: Matrix Scientific

030937-500MG
101831-138EA 290.77 USD
101831-138
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
3-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine

Hazard: Irritant

Formula: C₁₀H₁₄F₃N₃
MDL Number: MFCD03419763
CAS Number: 925580-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR