1-[3-(Difluoromethoxy)phenyl]ethanone

Supplier: Matrix Scientific

Synonyms: 3'-(Difluoromethoxy)acetophenone1-(3-(Difluoromethoxy)phenyl)ethanone

030936-500MG
101831-136EA 70.77 USD
101831-136
1-[3-(Difluoromethoxy)phenyl]ethanone
1-[3-(Difluoromethoxy)phenyl]ethanone

Hazard: Irritant

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
MDL Number: MFCD00236220
CAS Number: 101975-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR