4-Azepan-1-ylaniline dihydrochloride

Supplier: Matrix Scientific

030933-500MG
101831-130EA 290.77 USD
101831-130
4-Azepan-1-ylaniline dihydrochloride
4-Azepan-1-ylaniline dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₀Cl₂N₂
MDL Number: MFCD09431467
CAS Number: 57356-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR