1-[1-(4-Methoxyphenyl)-1H-pyrazol-4-yl]ethanamine

Supplier: Matrix Scientific

030928-500MG
101831-120EA 461.54 USD
101831-120
1-[1-(4-Methoxyphenyl)-1H-pyrazol-4-yl]ethanamine
1-[1-(4-Methoxyphenyl)-1H-pyrazol-4-yl]ethanamine

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O
MDL Number: MFCD08691420
CAS Number: 1049792-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR