1-[1-(4-Chlorophenyl)-1H-pyrazol-4-yl]ethanone

Supplier: Matrix Scientific

030924-500MG
101831-112EA 290.77 USD
101831-112
1-[1-(4-Chlorophenyl)-1H-pyrazol-4-yl]ethanone
1-[1-(4-Chlorophenyl)-1H-pyrazol-4-yl]ethanone

Hazard: Irritant

Formula: C₁₁H₉ClN₂O
MDL Number: MFCD08691477
CAS Number: 925580-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR