3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-(3-hydroxypropyl)pyrazole

030919-500MG
101831-102EA 444.62 USD
101831-102
3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol
3-(4-Bromo-1H-pyrazol-1-yl)propan-1-ol

Hazard: Irritant

Formula: C₆H₉BrN₂O
MW: 205.06 g/mol
MDL Number: MFCD06804869
CAS Number: 925180-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR