4-Bromo-1,3-dimethyl-1H-pyrazole ≥95%
Supplier: Matrix Scientific
030917-5G
030917-1G
101831-096EA
35.38
USD
101831-096
101831-098
4-Bromo-1,3-dimethyl-1H-pyrazole ≥95%
4-Bromo-1,3-dimethyl-1H-pyrazole
Hazard: Irritant
Formula:
C₅H₇BrN₂ MW: 175.0265 g/mol |
MDL Number:
MFCD00985488 CAS Number: 5775-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...