3-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)propan-1-ol

Supplier: Matrix Scientific

030916-500MG
101831-094EA 233.85 USD
101831-094
3-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)propan-1-ol
3-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)propan-1-ol

Hazard: Irritant

Formula: C₈H₁₃BrN₂O
MW: 233.11 g/mol
MDL Number: MFCD06804890
CAS Number: 925607-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR