2-(4-Bromo-1H-pyrazol-1-yl)ethanol

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-(2-hydroxyethyl)pyrazole

030915-500MG
101831-092EA 80 USD
101831-092
2-(4-Bromo-1H-pyrazol-1-yl)ethanol
2-(4-Bromo-1H-pyrazol-1-yl)ethanol

Hazard: Irritant

Formula: C₅H₇BrN₂O
MW: 191.03 g/mol
MDL Number: MFCD06804929
CAS Number: 214614-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR