2-(4-Bromo-1H-pyrazol-1-yl)propanoic acid

Supplier: Matrix Scientific

030913-500MG
101831-088EA 233.85 USD
101831-088
2-(4-Bromo-1H-pyrazol-1-yl)propanoic acid
2-(4-Bromo-1H-pyrazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₆H₇BrN₂O₂
MDL Number: MFCD04969759
CAS Number: 51292-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR