2-(4-Bromo-3-methyl-1H-pyrazol-1-yl)propanoic acid

Supplier: Matrix Scientific

030912-500MG
101831-086EA 290.77 USD
101831-086
2-(4-Bromo-3-methyl-1H-pyrazol-1-yl)propanoic acid
2-(4-Bromo-3-methyl-1H-pyrazol-1-yl)propanoic acid

Hazard: Irritant

Formula: C₇H₉BrN₂O₂
MDL Number: MFCD03419632
CAS Number: 925200-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR