3-Isoxazol-5-ylaniline

Supplier: Matrix Scientific

Synonyms: 3-(Isoxazol-5-yl)aniline

030901-500MG
101831-060EA 700 USD
101831-060
3-Isoxazol-5-ylaniline
3-Isoxazol-5-ylaniline

Hazard: Irritant

Formula: C₉H₈N₂O
MW: 160.18 g/mol
MDL Number: MFCD05667181
CAS Number: 832740-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR