Methyl-2-[(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]benzoate
Supplier: Matrix Scientific
030896-500MG
101831-050EA
193.85
USD
101831-050
Methyl-2-[(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]benzoate
Methyl-2-[(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)methyl]benzoate
Hazard: Irritant
Formula:
C₁₄H₁₅N₃O₄ |
MDL Number:
MFCD04969146 CAS Number: 925634-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...