1-[(2,4-Dimethylphenoxy)methyl]-1H-pyrazol-4-amine hydrochloride

Supplier: Matrix Scientific

030895-500MG
101831-048EA 321.54 USD
101831-048
1-[(2,4-Dimethylphenoxy)methyl]-1H-pyrazol-4-amine hydrochloride
1-[(2,4-Dimethylphenoxy)methyl]-1H-pyrazol-4-amine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O
MDL Number: MFCD09431206
CAS Number: 1049764-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR