Ethyl pyrazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 1H-pyrazole-3-carboxylate
, Pyrazole-3-carboxylic Acid Ethyl Ester
030893-25G
030893-5G
101831-042EA
27.69
USD
101831-042
101831-044
Ethyl pyrazole-3-carboxylate ≥95%
Ethyl pyrazole-3-carboxylate
Purity: 95+%
CAS: 5932-27-4
MDL Number: MFCD00159643
Molecular Formula: C6H8N2O2
Molecular Weight: 140,14
Hazard: Irritant
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol |
MDL Number:
MFCD00159643 CAS Number: 5932-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...