5-(4-Chlorobenzyl)-1H-tetrazole

Supplier: Matrix Scientific

030892-500MG
101831-038EA 193.85 USD
101831-038
5-(4-Chlorobenzyl)-1H-tetrazole
5-(4-Chlorobenzyl)-1H-tetrazole

Hazard: Irritant

Formula: C₈H₇ClN₄
MDL Number: MFCD00099429
CAS Number: 14064-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR