Methyl-5-(aminosulfonyl)-2,4-dichlorobenzoate

Supplier: Matrix Scientific

030891-500MG
101831-036EA 146.15 USD
101831-036
Methyl-5-(aminosulfonyl)-2,4-dichlorobenzoate
Methyl-5-(aminosulfonyl)-2,4-dichlorobenzoate

Hazard: Irritant

Formula: C₈H₇Cl₂NO₄S
MDL Number: MFCD00099226
CAS Number: 5046-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR