Methyl-5-(chloromethyl)-7-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Supplier: Matrix Scientific
030888-500MG
101831-030EA
193.85
USD
101831-030
Methyl-5-(chloromethyl)-7-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Methyl-5-(chloromethyl)-7-hydroxypyrazolo[1,5-a]pyrimidine-3-carboxylate
Hazard: Irritant
Formula:
C₉H₈ClN₃O₃ |
MDL Number:
MFCD03450753 CAS Number: 656818-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...