[3-(Difluoromethyl)-5-methyl-4-nitro-1H-pyrazol-1-yl]acetic acid

Supplier: Matrix Scientific

030881-500MG
101831-016EA 290.77 USD
101831-016
[3-(Difluoromethyl)-5-methyl-4-nitro-1H-pyrazol-1-yl]acetic acid
[3-(Difluoromethyl)-5-methyl-4-nitro-1H-pyrazol-1-yl]acetic acid

Hazard: Irritant

Formula: C₇H₇F₂N₃O₄
MDL Number: MFCD04969938
CAS Number: 925607-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR