1-Benzyl-3,5-dimethyl-4-nitro-1H-pyrazole

Supplier: Matrix Scientific

030878-500MG
101831-010EA 193.85 USD
101831-010
1-Benzyl-3,5-dimethyl-4-nitro-1H-pyrazole
1-Benzyl-3,5-dimethyl-4-nitro-1H-pyrazole

Hazard: Irritant

Formula: C₁₂H₁₃N₃O₂
MDL Number: MFCD00546042
CAS Number: 100193-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR