(2E)-3-(4-Ethoxyphenyl)-1-(1-ethyl-3-methyl-1H-pyrazol-4-yl)prop-2-en-1-one
Supplier: Matrix Scientific
030876-500MG
101831-006EA
193.85
USD
101831-006
(2E)-3-(4-Ethoxyphenyl)-1-(1-ethyl-3-methyl-1H-pyrazol-4-yl)prop-2-en-1-one
(2E)-3-(4-Ethoxyphenyl)-1-(1-ethyl-3-methyl-1H-pyrazol-4-yl)prop-2-en-1-one
Hazard: Irritant
Formula:
C₁₇H₂₀N₂O₂ |
MDL Number:
MFCD04045503 CAS Number: 955563-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...