1-[(2-Chlorophenoxy)methyl]-1H-pyrazole-5-carboxylic acid

Supplier: Matrix Scientific

030874-500MG
101831-002EA 193.85 USD
101831-002
1-[(2-Chlorophenoxy)methyl]-1H-pyrazole-5-carboxylic acid
1-[(2-Chlorophenoxy)methyl]-1H-pyrazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₃
MDL Number: MFCD04969815
CAS Number: 925606-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR