1-[(1-Methyl-1H-pyrazol-4-yl)sulfonyl]piperidine-4-carboxylic acid

Supplier: Matrix Scientific

030870-500MG
101830-994EA 193.85 USD
101830-994
1-[(1-Methyl-1H-pyrazol-4-yl)sulfonyl]piperidine-4-carboxylic acid
1-[(1-Methyl-1H-pyrazol-4-yl)sulfonyl]piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₄S
MDL Number: MFCD07400409
CAS Number: 925178-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR