1-(1-Ethyl-5-methyl-1H-pyrazol-3-yl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

030868-500MG
101830-990EA 336.92 USD
101830-990
1-(1-Ethyl-5-methyl-1H-pyrazol-3-yl)-5-oxopyrrolidine-3-carboxylic acid
1-(1-Ethyl-5-methyl-1H-pyrazol-3-yl)-5-oxopyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₃
MDL Number: MFCD09431135
CAS Number: 925199-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR