Methyl-5-(1,5-dimethyl-1H-pyrazol-4-yl)isoxazole-3-carboxylate

Supplier: Matrix Scientific

030867-500MG
101830-988EA 290.77 USD
101830-988
Methyl-5-(1,5-dimethyl-1H-pyrazol-4-yl)isoxazole-3-carboxylate
Methyl-5-(1,5-dimethyl-1H-pyrazol-4-yl)isoxazole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₁N₃O₃
MDL Number: MFCD04971121
CAS Number: 925178-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR