1-Benzyl-2,3-dimethyl-1H-indole-5-carboxylic acid

Supplier: Matrix Scientific

030866-500MG
101830-986EA 304.62 USD
101830-986
1-Benzyl-2,3-dimethyl-1H-indole-5-carboxylic acid
1-Benzyl-2,3-dimethyl-1H-indole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₈H₁₇NO₂
MDL Number: MFCD00419414
CAS Number: 313498-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR