5-Bromo-N-ethylthiophene-2-carboxamide

Supplier: Matrix Scientific

030860-500MG
101830-974EA 146.15 USD
101830-974
5-Bromo-N-ethylthiophene-2-carboxamide
5-Bromo-N-ethylthiophene-2-carboxamide

Hazard: Irritant

Formula: C₇H₈BrNOS
MDL Number: MFCD08593585
CAS Number: 908518-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR