5-Bromo-N-isopropylthiophene-2-carboxamide

Supplier: Matrix Scientific

030855-500MG
101830-964EA 146.15 USD
101830-964
5-Bromo-N-isopropylthiophene-2-carboxamide
5-Bromo-N-isopropylthiophene-2-carboxamide

Hazard: Irritant

Formula: C₈H₁₀BrNOS
MDL Number: MFCD08593344
CAS Number: 908494-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR