3-Chloro-N-2-naphthylpropanamide

Supplier: Matrix Scientific

030851-500MG
101830-956EA 146.15 USD
101830-956
3-Chloro-N-2-naphthylpropanamide
3-Chloro-N-2-naphthylpropanamide

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
MDL Number: MFCD01341947
CAS Number: 183111-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR