3-Chloro-N-(3-ethylphenyl)propanamide

Supplier: Matrix Scientific

030849-500MG
101830-952EA 146.15 USD
101830-952
3-Chloro-N-(3-ethylphenyl)propanamide
3-Chloro-N-(3-ethylphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₄ClNO
MDL Number: MFCD01337291
CAS Number: 573997-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR