N-(2-Acetylphenyl)-3-chloropropanamide

Supplier: Matrix Scientific

030847-500MG
101830-948EA 146.15 USD
101830-948
N-(2-Acetylphenyl)-3-chloropropanamide
N-(2-Acetylphenyl)-3-chloropropanamide

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₂
MDL Number: MFCD08593323
CAS Number: 18014-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR