1-(3-Methyl-1-adamantyl)piperazine

Supplier: Matrix Scientific

030846-500MG
101830-946EA 290.77 USD
101830-946
1-(3-Methyl-1-adamantyl)piperazine
1-(3-Methyl-1-adamantyl)piperazine

Hazard: Irritant

Formula: C₁₅H₂₆N₂
MDL Number: MFCD08569399
CAS Number: 906789-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR