4-Cyclopropyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030841-500MG
101830-936EA 193.85 USD
101830-936
4-Cyclopropyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol
4-Cyclopropyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₂H₂₀N₄O₂S₂
MDL Number: MFCD08569924
CAS Number: 932830-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR