4-Benzyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030840-500MG
101830-934EA 193.85 USD
101830-934
4-Benzyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol
4-Benzyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₆H₂₂N₄O₂S₂
MDL Number: MFCD08569923
CAS Number: 932875-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR