5-[1-(Ethylsulfonyl)piperidin-3-yl]-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030839-500MG
101830-932EA 253.85 USD
101830-932
5-[1-(Ethylsulfonyl)piperidin-3-yl]-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol
5-[1-(Ethylsulfonyl)piperidin-3-yl]-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₄H₂₄N₄O₃S₂
MDL Number: MFCD08569922
CAS Number: 932918-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR