4-(3,4-Dimethoxyphenethyl)-5-(1-(ethylsulfonyl)piperidin-3-yl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030838-500MG
101830-930EA 253.85 USD
101830-930
4-(3,4-Dimethoxyphenethyl)-5-(1-(ethylsulfonyl)piperidin-3-yl)-4H-1,2,4-triazole-3-thiol
4-(3,4-Dimethoxyphenethyl)-5-(1-(ethylsulfonyl)piperidin-3-yl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₉H₂₈N₄O₄S₂
MDL Number: MFCD08569921
CAS Number: 932807-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR