5-[1-(Ethylsulfonyl)piperidin-3-yl]-4-propyl-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030835-500MG
101830-924EA 193.85 USD
101830-924
5-[1-(Ethylsulfonyl)piperidin-3-yl]-4-propyl-4H-1,2,4-triazole-3-thiol
5-[1-(Ethylsulfonyl)piperidin-3-yl]-4-propyl-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₂H₂₂N₄O₂S₂
MDL Number: MFCD08569918
CAS Number: 932918-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR