5-(1-(Ethylsulfonyl)piperidin-3-yl)-4-(4-methoxyphenethyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030834-500MG
101830-922EA 193.85 USD
101830-922
5-(1-(Ethylsulfonyl)piperidin-3-yl)-4-(4-methoxyphenethyl)-4H-1,2,4-triazole-3-thiol
5-(1-(Ethylsulfonyl)piperidin-3-yl)-4-(4-methoxyphenethyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₈H₂₆N₄O₃S₂
MW: 410.56 g/mol
MDL Number: MFCD08569917
CAS Number: 932807-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR