4-Butyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

030829-500MG
101830-912EA 173.85 USD
101830-912
4-Butyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol
4-Butyl-5-[1-(ethylsulfonyl)piperidin-3-yl]-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₁₃H₂₄N₄O₂S₂
MDL Number: MFCD08569902
CAS Number: 932830-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR