2-(3-Bromophenoxy)propanamide

Supplier: Matrix Scientific

030823-500MG
101830-900EA 146.15 USD
101830-900
2-(3-Bromophenoxy)propanamide
2-(3-Bromophenoxy)propanamide

Hazard: Irritant

Formula: C₉H₁₀BrNO₂
MDL Number: MFCD08569898
CAS Number: 915923-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR