2-Chloro-N-ethylnicotinamide

Supplier: Matrix Scientific

030822-500MG
101830-898EA 164.62 USD
101830-898
2-Chloro-N-ethylnicotinamide
2-Chloro-N-ethylnicotinamide

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD01358076
CAS Number: 52943-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR