3-[(3,3-Dimethylbutanoyl)amino]-4-methylbenzoic acid

Supplier: Matrix Scientific

030818-500MG
101830-890EA 146.15 USD
101830-890
3-[(3,3-Dimethylbutanoyl)amino]-4-methylbenzoic acid
3-[(3,3-Dimethylbutanoyl)amino]-4-methylbenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₃
MDL Number: MFCD08569895
CAS Number: 915921-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR