3-[(3,3-Dimethylbutanoyl)amino]benzoic acid

Supplier: Matrix Scientific

030817-500MG
101830-888EA 146.15 USD
101830-888
3-[(3,3-Dimethylbutanoyl)amino]benzoic acid
3-[(3,3-Dimethylbutanoyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD08569894
CAS Number: 915923-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR