3-[(Cyclobutylcarbonyl)amino]-4-methylbenzoic acid

Supplier: Matrix Scientific

030816-500MG
101830-886EA 146.15 USD
101830-886
3-[(Cyclobutylcarbonyl)amino]-4-methylbenzoic acid
3-[(Cyclobutylcarbonyl)amino]-4-methylbenzoic acid

Hazard: Irritant

Formula: C₁₃H₁₅NO₃
MDL Number: MFCD08569893
CAS Number: 915921-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR