3-Chloro-N-(thien-2-ylmethyl)propanamide

Supplier: Matrix Scientific

030815-500MG
101830-884EA 307.69 USD
101830-884
3-Chloro-N-(thien-2-ylmethyl)propanamide
3-Chloro-N-(thien-2-ylmethyl)propanamide

Hazard: Irritant

Formula: C₈H₁₀ClNOS
MDL Number: MFCD03394727
CAS Number: 21403-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR