3-(Isobutyrylamino)benzoic acid

Supplier: Matrix Scientific

030809-500MG
101830-872EA 82.38 USD
101830-872
3-(Isobutyrylamino)benzoic acid
3-(Isobutyrylamino)benzoic acid

CAS: 28533-44-0
MDL Number: MFCD00227316
Molecular Formula: C11H13NO3
Molecular Weight: 207,23
Hazard: Irritant

Formula: C₁₁H₁₃NO₃
MW: 207,23 g/mol
MDL Number: MFCD00227316
CAS Number: 28533-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR