3-[(Cyclopropylcarbonyl)amino]-4-methylbenzoic acid

Supplier: Matrix Scientific

030808-500MG
101830-870EA 146.15 USD
101830-870
3-[(Cyclopropylcarbonyl)amino]-4-methylbenzoic acid
3-[(Cyclopropylcarbonyl)amino]-4-methylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD08569890
CAS Number: 915923-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR