Methyl-3-[(3-chloropropanoyl)amino]thiophene-2-carboxylate
Supplier: Matrix Scientific
030807-500MG
101830-868EA
193.85
USD
101830-868
Methyl-3-[(3-chloropropanoyl)amino]thiophene-2-carboxylate
Methyl-3-[(3-chloropropanoyl)amino]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₉H₁₀ClNO₃S |
MDL Number:
MFCD01340734 CAS Number: 549478-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...