3-Bromo-5-methoxy-4-(2-oxo-2-phenylethoxy)benzaldehyde

Supplier: Matrix Scientific

030793-500MG
101830-840EA 387.69 USD
101830-840
3-Bromo-5-methoxy-4-(2-oxo-2-phenylethoxy)benzaldehyde
3-Bromo-5-methoxy-4-(2-oxo-2-phenylethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₆H₁₃BrO₄
MW: 349.19 g/mol
MDL Number: MFCD05995935
CAS Number: 712348-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR