3-(2-Oxo-2-phenylethoxy)benzaldehyde

Supplier: Matrix Scientific

030790-500MG
101830-834EA 387.69 USD
101830-834
3-(2-Oxo-2-phenylethoxy)benzaldehyde
3-(2-Oxo-2-phenylethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₅H₁₂O₃
MDL Number: MFCD09971998
CAS Number: 956576-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR